Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
TACR1
Transcript:
Assay Name:
Assay ID
CRISPR608249_CR
Size
2 nmol Synthetic crRNA
2 nmol Synthetic crRNA
Availability
Made To Order
Catalog #
A35509
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Panther Classification:
Molecular Function -
G-protein coupled receptor
receptor
Gene Ontology Categories:
Function(s)
Process(es)
aggressive behavior
acute inflammatory response
positive regulation of leukocyte migration
angiotensin-mediated drinking behavior
inflammatory response
cell surface receptor signaling pathway
phospholipase C-activating G-protein coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
tachykinin receptor signaling pathway
chemical synaptic transmission
long-term memory
associative learning
response to heat
detection of abiotic stimulus
response to ozone
positive regulation of epithelial cell migration
response to auditory stimulus
regulation of smooth muscle cell migration
sensory perception of pain
positive regulation of synaptic transmission, cholinergic
positive regulation of synaptic transmission, GABAergic
response to estradiol
response to progesterone
response to nicotine
operant conditioning
positive regulation of renal sodium excretion
sperm ejaculation
eating behavior
positive regulation of vascular permeability
response to morphine
response to ethanol
positive regulation of action potential
positive regulation of blood pressure
positive regulation of ossification
positive regulation of vasoconstriction
positive regulation of smooth muscle contraction
positive regulation of saliva secretion
positive regulation of hormone secretion
behavioral response to pain
regulation of smooth muscle cell proliferation
positive regulation of lymphocyte proliferation
positive regulation of epithelial cell proliferation
positive regulation of stress fiber assembly
response to electrical stimulus
smooth muscle contraction involved in micturition
positive regulation of uterine smooth muscle contraction
tachykinin receptor activity
protein binding
substance P receptor activity
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-9fjjm:80/10.239.165.64:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0