Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
ACVR1
Transcript:
Assay Name:
Assay ID
CRISPR686970_LV
Size
200 µL,1x10⁸ TU/mL
200 µL,1x10⁸ TU/mL
Availability
Made To Order
Catalog #
A32042
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Panther Classification:
Molecular Function -
TGF-beta receptor
cytokine receptor
kinase
protein kinase
protein kinase receptor
receptor
serine/threonine protein kinase receptor
transferase
Gene Ontology Categories:
Function(s)
Process(es)
G1/S transition of mitotic cell cycle
patterning of blood vessels
in utero embryonic development
gastrulation with mouth forming second
mesoderm formation
neural crest cell migration
acute inflammatory response
embryonic heart tube morphogenesis
outflow tract septum morphogenesis
atrioventricular valve morphogenesis
mitral valve morphogenesis
endocardial cushion morphogenesis
endocardial cushion fusion
atrial septum primum morphogenesis
protein phosphorylation
transforming growth factor beta receptor signaling pathway
germ cell development
determination of left/right symmetry
negative regulation of signal transduction
positive regulation of pathway-restricted SMAD protein phosphorylation
peptidyl-threonine phosphorylation
signal transduction by protein phosphorylation
regulation of ossification
positive regulation of cell migration
positive regulation of bone mineralization
BMP signaling pathway
activin receptor signaling pathway
negative regulation of activin receptor signaling pathway
positive regulation of osteoblast differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
smooth muscle cell differentiation
pharyngeal system development
pathway-restricted SMAD protein phosphorylation
ventricular septum morphogenesis
cardiac muscle cell fate commitment
BMP signaling pathway involved in heart development
endocardial cushion cell fate commitment
cellular response to glucocorticoid stimulus
cellular response to BMP stimulus
positive regulation of epithelial to mesenchymal transition involved in endocardial cushion formation
positive regulation of determination of dorsal identity
negative regulation of extrinsic apoptotic signaling pathway
protein kinase activity
protein serine/threonine kinase activity
transmembrane receptor protein serine/threonine kinase activity
receptor signaling protein serine/threonine kinase activity
transforming growth factor beta receptor activity, type I
protein binding
ATP binding
activin receptor activity, type I
peptide hormone binding
protein homodimerization activity
SMAD binding
metal ion binding
activin binding
transforming growth factor beta binding
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-f6n66:80/10.239.161.116:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0