Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
DRD3
Transcript:
Assay Name:
Assay ID
CRISPR779749_CR
Size
2 nmol Synthetic crRNA
2 nmol Synthetic crRNA
Availability
Made To Order
Catalog #
A35509
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Panther Classification:
Molecular Function -
G-protein coupled receptor
receptor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
synaptic transmission, dopaminergic
response to amphetamine
regulation of blood volume by renin-angiotensin
G-protein coupled receptor internalization
cellular calcium ion homeostasis
G-protein coupled receptor signaling pathway
adenylate cyclase-activating dopamine receptor signaling pathway
negative regulation of adenylate cyclase activity
adenylate cyclase-inhibiting dopamine receptor signaling pathway
learning or memory
learning
locomotory behavior
positive regulation of cell proliferation
visual learning
regulation of dopamine secretion
regulation of lipid metabolic process
regulation of cAMP metabolic process
negative regulation of sodium:proton antiporter activity
positive regulation of cytokinesis
circadian regulation of gene expression
response to histamine
social behavior
gastric emptying
positive regulation of renal sodium excretion
regulation of multicellular organism growth
response to cocaine
dopamine metabolic process
response to drug
response to morphine
regulation of circadian sleep/wake cycle, sleep
response to ethanol
negative regulation of blood pressure
positive regulation of mitotic nuclear division
positive regulation of transcription from RNA polymerase II promoter
acid secretion
behavioral response to cocaine
negative regulation of oligodendrocyte differentiation
arachidonic acid secretion
negative regulation of protein secretion
musculoskeletal movement, spinal reflex action
regulation of dopamine uptake involved in synaptic transmission
negative regulation of protein kinase B signaling
prepulse inhibition
negative regulation of dopamine receptor signaling pathway
positive regulation of dopamine receptor signaling pathway
regulation of locomotion involved in locomotory behavior
dopamine neurotransmitter receptor activity, coupled via Gs
dopamine neurotransmitter receptor activity, coupled via Gi/Go
protein binding
drug binding
protein domain specific binding
D1 dopamine receptor binding
dopamine binding
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-gk48b:80/10.239.167.222:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0