Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
FLT3
Transcript:
Assay Name:
Assay ID
CRISPR865936_CR
Size
2 nmol Synthetic crRNA
2 nmol Synthetic crRNA
Availability
Made To Order
Catalog #
A35509
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
leukocyte homeostasis
myeloid progenitor cell differentiation
pro-B cell differentiation
transmembrane receptor protein tyrosine kinase signaling pathway
positive regulation of cell proliferation
response to organonitrogen compound
positive regulation of phosphatidylinositol 3-kinase signaling
peptidyl-tyrosine phosphorylation
cytokine-mediated signaling pathway
hemopoiesis
B cell differentiation
organ regeneration
common myeloid progenitor cell proliferation
vascular endothelial growth factor signaling pathway
positive regulation of tyrosine phosphorylation of STAT protein
regulation of apoptotic process
positive regulation of MAP kinase activity
positive regulation of MAPK cascade
positive regulation of phosphatidylinositol 3-kinase activity
lymphocyte proliferation
protein autophosphorylation
cellular response to cytokine stimulus
cellular response to glucocorticoid stimulus
dendritic cell differentiation
transmembrane receptor protein tyrosine kinase activity
cytokine receptor activity
vascular endothelial growth factor-activated receptor activity
protein binding
ATP binding
protein complex binding
glucocorticoid receptor binding
protein homodimerization activity
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-9fjjm:80/10.239.165.64:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0