Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
AXL
Transcript:
Assay Name:
Assay ID
CRISPR932417_CR
Size
2 nmol Synthetic crRNA
2 nmol Synthetic crRNA
Availability
Made To Order
Catalog #
A35509
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
neuron migration
natural killer cell differentiation
positive regulation of cytokine-mediated signaling pathway
blood vessel remodeling
phagocytosis
inflammatory response
signal transduction
spermatogenesis
peptidyl-tyrosine phosphorylation
forebrain cell migration
platelet activation
cellular response to extracellular stimulus
negative regulation of interferon-gamma production
negative regulation of tumor necrosis factor production
positive regulation of natural killer cell differentiation
secretion by cell
erythrocyte homeostasis
substrate adhesion-dependent cell spreading
cellular response to interferon-alpha
ovulation cycle
apoptotic cell clearance
protein kinase B signaling
negative regulation of neuron apoptotic process
innate immune response
viral entry into host cell
vascular endothelial growth factor receptor signaling pathway
cell maturation
positive regulation of pinocytosis
negative regulation of lymphocyte activation
positive regulation of protein kinase B signaling
vagina development
cellular response to lipopolysaccharide
dendritic cell differentiation
negative regulation of dendritic cell apoptotic process
virus receptor activity
phosphatidylserine binding
protein tyrosine kinase activity
transmembrane receptor protein tyrosine kinase activity
protein binding
ATP binding
protein heterodimerization activity
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-c74kb:80/10.239.167.51:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0