Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
CREBBP
Transcript:
Assay Name:
Assay ID
CRISPR974865_CR
Size
2 nmol Synthetic crRNA
2 nmol Synthetic crRNA
Availability
Made To Order
Catalog #
A35509
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA binding protein
acetyltransferase
chromatin/chromatin-binding protein
nucleic acid binding
transcription cofactor
transcription factor
transferase
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
response to hypoxia
stimulatory C-type lectin receptor signaling pathway
regulation of transcription, DNA-templated
transcription initiation from RNA polymerase II promoter
protein complex assembly
protein acetylation
signal transduction
Notch signaling pathway
cell proliferation
regulation of smoothened signaling pathway
viral process
histone acetylation
N-terminal peptidyl-lysine acetylation
positive regulation of type I interferon production
cellular response to UV
homeostatic process
embryonic digit morphogenesis
regulation of apoptotic process
cellular lipid metabolic process
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
rhythmic process
positive regulation of cell adhesion molecule production
regulation of transcription from RNA polymerase II promoter in response to hypoxia
response to interleukin-1
cell-cell adhesion
regulation of cellular response to heat
positive regulation of G1/S transition of mitotic cell cycle
positive regulation of NIK/NF-kappaB signaling
beta-catenin-TCF complex assembly
core promoter proximal region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
RNA polymerase II transcription factor binding
RNA polymerase II activating transcription factor binding
RNA polymerase II transcription coactivator activity
transcriptional repressor activity, RNA polymerase II transcription factor binding
p53 binding
chromatin binding
damaged DNA binding
transcription factor activity, sequence-specific DNA binding
transcription coactivator activity
histone acetyltransferase activity
signal transducer activity
protein binding
transcription factor binding
zinc ion binding
acetyltransferase activity
peptide N-acetyltransferase activity
peroxisome proliferator activated receptor binding
MRF binding
SMAD binding
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-jgtjs:80/10.239.162.121:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0