Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfkbib | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
GenBank mRNA | AK010218.1 | - | ||||
AK038631.1 | - | |||||
AK156731.1 | - | |||||
AK182221.1 | - | |||||
AK202557.1 | - | |||||
AK213461.1 | - | |||||
AY176058.1 | - | |||||
AY176059.1 | - | |||||
BC021938.1 | - | 5136809 | ||||
BM899229.1 | - | |||||
U19799.1 | - |