Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC1A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001195728.2 | NP_001182657.1 | ||||
NM_001252652.1 | NP_001239581.1 | |||||
NM_004171.3 | NP_004162.2 | |||||
XM_005253067.1 | XP_005253124.1 | |||||
XM_011520285.2 | XP_011518587.1 | |||||
XM_017018136.1 | XP_016873625.1 | |||||
XM_017018137.1 | XP_016873626.1 | |||||
XM_017018138.1 | XP_016873627.1 | |||||
XM_017018139.1 | XP_016873628.1 | |||||
GenBank mRNA | AB209444.1 | - | ||||
AF131756.1 | - | |||||
AK054855.1 | - | |||||
AK057674.1 | - | |||||
AK090810.1 | - | |||||
AK122843.1 | - | |||||
AK298769.1 | - | |||||
AL157452.1 | - | |||||
AY066021.1 | - | |||||
BC031313.2 | - | |||||
BC047052.1 | - | |||||
BC132768.1 | - | 40146790 | ||||
BX509751.1 | - | |||||
BX538155.1 | - | |||||
BX648750.1 | - | |||||
CR626999.1 | - | |||||
CR627009.1 | - | |||||
CR627022.1 | - | |||||
CR749842.1 | - | |||||
D85884.1 | - | |||||
DA158200.1 | - | |||||
DA215039.1 | - | |||||
DB465494.2 | - | |||||
ENST00000278379.0 | - | |||||
KP966086.1 | - | |||||
S83374.1 | - | |||||
S83375.1 | - | |||||
U01824.1 | - | |||||
U03505.1 | - | |||||
Z32517.1 | - |