Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_174899.4 | NP_777559.3 | ||||
XM_005246317.1 | XP_005246374.1 | |||||
GenBank mRNA | AK094911.1 | - | ||||
AK123085.1 | - | |||||
AK289655.1 | - | |||||
AK309702.1 | - | |||||
BC017869.1 | - | |||||
BC022538.1 | - | 4817261 | ||||
BC033935.1 | - | 5296808 | ||||
BF674223.1 | - | |||||
BM997950.1 | - | |||||
ENST00000283946.0 | - |