Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itgb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_008404.4 | NP_032430.2 | ||||
GenBank mRNA | AK088435.1 | - | ||||
AK136502.1 | - | |||||
AK161236.1 | - | |||||
AK169587.1 | - | |||||
BC145644.1 | - | 40130947 | ||||
M31039.1 | - | |||||
X14951.1 | - |