Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_010612.2 | NP_034742.2 | ||||
GenBank mRNA | AK031739.1 | - | ||||
AK054510.1 | - | |||||
AK141938.1 | - | |||||
BC020530.1 | - | 4238984 | ||||
EU884114.1 | - | |||||
S53103.1 | - | |||||
X59397.1 | - | |||||
X70842.1 | - |