Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_007922.5 | NP_031948.4 | ||||
GenBank mRNA | AK082260.1 | - | ||||
AK132354.1 | - | |||||
AK196053.1 | - | |||||
AK216408.1 | - | |||||
BC005706.1 | - | |||||
BC054474.1 | - | 30045506 | ||||
BY728635.1 | - | |||||
CV561936.1 | - | |||||
D26066.1 | - | |||||
FN434115.1 | - | |||||
X87257.1 | - | |||||
Z36939.1 | - |