Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mertk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_008587.1 | NP_032613.1 | ||||
GenBank mRNA | AK029009.1 | - | ||||
AK030158.1 | - | |||||
AK079252.1 | - | |||||
L11625.1 | - | |||||
U21301.1 | - |