Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgf16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_030614.2 | NP_085117.2 | ||||
GenBank mRNA | AB049219.1 | - | ||||
AF292104.1 | - | |||||
BB342405.2 | - | |||||
BC132641.1 | - | 40130918 | ||||
BC137993.1 | - | 8861012 |