Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnase1l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_007870.3 | NP_031896.3 | ||||
GenBank mRNA | AF047355.1 | - | ||||
AK156109.1 | - | |||||
AK164072.1 | - | |||||
AK171843.1 | - | |||||
AK172254.1 | - | |||||
BC012671.1 | - | 4160709 | ||||
U76110.1 | - |