Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_028384.1 | NP_082660.1 | ||||
GenBank mRNA | AK013632.1 | - | ||||
AK038868.1 | - | |||||
AK158659.1 | - | |||||
AW911821.1 | - | |||||
BB762565.1 | - | |||||
BB857029.1 | - | |||||
BC147354.1 | - | 9056407 | ||||
BG800529.1 | - | |||||
BM949737.1 | - | |||||
BY082969.1 | - | |||||
BY296983.1 | - |