Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lsd-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
RefSeq | NM_001042811.2 | NP_001036276.1 | ||||
NM_001042812.2 | NP_001036277.2 | |||||
NM_001298332.1 | NP_001285261.1 | |||||
NM_001298333.1 | NP_001285262.1 | |||||
NM_132768.3 | NP_572996.1 | |||||
GenBank mRNA | AY060242.1 | - |