Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128714.4 | NP_180715.1 | 1 - 2 | 194 | 73 | |
GenBank mRNA | AY044330.1 | - | 1 - 2 | 46 | 73 | |
AY058187.1 | - | 1 - 2 | 149 | 73 | ||
AY086518.1 | - | 1 - 2 | 133 | 73 | ||
AY098982.1 | - | 1 - 2 | 46 | 73 | ||
U94495.1 | - | 1 - 2 | 71 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |