Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPS8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334443.1 | NP_001321199.1 | 3 - 4 | 2103 | 70 | |
NM_105697.4 | NP_177186.2 | 2 - 3 | 2264 | 70 | ||
GenBank mRNA | AK227167.1 | - | 2 - 3 | 2167 | 70 | |
KJ676991.1 | - | 1 - 2 | 1977 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |