Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: abts-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136304.4 | NP_001129776.1 | 4 - 5 | 965 | 86 | |
NM_001380825.1 | NP_001366789.1 | 4 - 5 | 488 | 86 | ||
GenBank mRNA | AY887903.1 | - | 4 - 5 | 510 | 86 | |
DQ178237.1 | - | 3 - 4 | 646 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |