Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rhr-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073560.6 | NP_505961.1 | 10 - 11 | 1317 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |