Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PVALB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003431505.3 | XP_003431553.1 | 4 - 5 | 363 | 85 | |
XM_538399.6 | XP_538399.2 | 4 - 5 | 442 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Canis familiaris Probe spans exons |