Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cfh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130409.2 | NP_569093.2 | 19 - 20 | 3195 | 102 | |
XM_008769531.1 | XP_008767753.1 | 18 - 19 | 3221 | 102 | ||
XM_008769532.1 | XP_008767754.1 | 10 - 11 | 1753 | 102 | ||
GenBank mRNA | AJ320522.1 | - | 3169 | 102 | ||
BC083174.1 | - | 1566 | 7096787 | 102 | ||
CF110979.1 | - | 3 - 4 | 551 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |