Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ficd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001010946.1 | NP_001010946.1 | 1 - 2 | 706 | 128 | |
XM_008769320.1 | XP_008767542.1 | 2 - 3 | 603 | 128 | ||
XM_008769321.1 | XP_008767543.1 | 2 - 3 | 533 | 128 | ||
GenBank mRNA | BC079197.1 | - | 1 - 2 | 706 | 7131463 | 128 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |