Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116904.6 | NP_192575.3 | 20 - 21 | 2981 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |