Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036849.2 | NP_001031926.1 | 12 - 13 | 1806 | 71 | |
NM_122214.4 | NP_197699.2 | 13 - 14 | 1983 | 71 | ||
GenBank mRNA | AK118129.1 | - | 13 - 14 | 1891 | 71 | |
AY518689.1 | - | 12 - 13 | 1762 | 71 | ||
BT008588.1 | - | 13 - 14 | 1852 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |