Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ISOC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101213.2 | NP_001094683.1 | 4 - 5 | 813 | 80 | |
GenBank mRNA | BC148130.1 | - | 4 - 5 | 814 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |