Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: serpine2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200184.1 | NP_956478.1 | 4 - 5 | 776 | 72 | |
GenBank mRNA | BC045498.1 | - | 4 - 5 | 776 | 72 | |
BC163961.1 | - | 3 - 4 | 718 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |