Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1200002994.mm.1 |
NM_029760.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200002994.mm.1 |
NM_029760.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200002994.mm.1 |
TC1200000422.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200002995.mm.1 |
NM_029760.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200002995.mm.1 |
NM_029760.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200002995.mm.1 |
TC1200000422.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200002997.mm.1 |
NM_029760.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200002997.mm.1 |
NM_029760.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200002997.mm.1 |
TC1200000422.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200002993.mm.1 |
NM_029760.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200002993.mm.1 |
NM_029760.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200002993.mm.1 |
TC1200000422.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200002996.mm.1 |
NM_029760.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200002996.mm.1 |
NM_029760.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200002996.mm.1 |
TC1200000422.mm.1 |
MTA TC |