Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dctn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026302.3 | NP_080578.1 | 12 | 1097 | ||
GenBank mRNA | AK016059.1 | - | 12 | 1151 | ||
AK034292.1 | - | 13 | 1155 | |||
AK141855.1 | - | 13 | 1120 | |||
AK160813.1 | - | 12 | 1103 | |||
AK163240.1 | - | 12 | 1119 | |||
AK168484.1 | - | 12 | 1103 | |||
BC006677.1 | - | 12 | 1060 | |||
BC034725.1 | - | 12 | 1097 | 3667031 |