Talk with Us
Episode 051
When Wheat Matters: Allergy, Celiac Disease, and Intolerances
Episode summary
Wheat allergy, celiac disease, and non-specific gluten intolerance all share overlapping gastrointestinal symptoms that can confound primary care diagnosis. This episode addresses the core diagnostic challenge of distinguishing IgE-mediated wheat allergy (including ω-5 gliadin–associated wheat-dependent exercise-induced anaphylaxis) from celiac disease and non-celiac gluten intolerance. We cover epidemiology, clinical presentations and timing of IgE reactions, component-resolved diagnostics, serologic algorithms for celiac disease, when to refer for biopsy, and management implications including gluten-free diet counseling and emergency planning for anaphylaxis. Practical takeaways emphasize improving diagnostic yield in primary care, appropriate use of specific IgE testing, and when specialist referral is necessary.
Episode resources
Find the resources mentioned in this episode
Lab Ordering Guide
Looking for allergy diagnostic codes from the labs you already use?
Episode transcript
Time stamps
00:00 — Episode Overview and Objectives
02:39 — Wheat Allergy Prevalence and Natural History
03:14 — IgE Symptoms and Cross Sensitization
03:54 — Omega 5 Gliadin and WDEIA
06:21 — Celiac Prevalence and Diagnostic Gap
08:11 — Celiac Pathophysiology and Gluten Free Diet
09:38 — Serologic Workup and Biopsy Guidance
12:28 — Non Celiac Gluten Sensitivity and Diagnostic Limitations
15:32 — IBS Differential and Alpha Gal
16:07 — Clinical Takeaways for Primary Care
References used in this episode
  • Armstrong, David, and Elena F. Verdú. “The Spectrum of Wheat Sensitivity: From Non-Celiac Gluten Sensitivity to Celiac Disease.” The Spectrum of Wheat Sensitivity, Association for Diagnostics and Laboratory Medicine (ADLM), 1 Aug. 2014, https://myadlm.org/cln/articles/2014/august/wheat-sensitivity
  • Sicherer SH, Warren CM, Dant C, Gupta RS, Nadeau KC. Food Allergy from Infancy Through Adulthood. J Allergy Clin Immunol Pract. 2020 Jun;8(6):1854-1864.
  • Gupta RS, Warren CM, Smith BM, et al. Prevalence and Severity of Food Allergies Among US Adults. JAMA Netw Open. 2019;2(1):e185630.
  • Nilsson N, Nilsson C, Ekoff H, Wieser-Pahr S, Borres MP, Valenta R, Hedlin G, Sjölander S. Grass-Allergic Children Frequently Show Asymptomatic Low-Level IgE Co-Sensitization and Cross-Reactivity to Wheat. Int Arch Allergy Immunol. 2018;177(2):135-144.
  • Kennard L, Thomas I, Rutkowski K, Azzu V, Yong PFK, Kasternow B, Hunter H, Cabdi NMO, Nakonechna A, Wagner A. A Multicenter Evaluation of Diagnosis and Management of Omega-5 Gliadin Allergy (Also Known as Wheat-Dependent Exercise-Induced Anaphylaxis) in 132 Adults. J Allergy Clin Immunol Pract. 2018 Nov-Dec;6(6):1892-1897.
  • Beyond Celiac. “Celiac Disease: Facts and Figures.” BeyondCeliac.org, 2025, https://www.beyondceliac.org/celiac-disease/facts-and-figures/. Accessed 3 Mar. 2026.
  • Du Y, Shan LF, Cao ZZ, Feng JC, Cheng Y. Prevalence of celiac disease in patients with Down syndrome: a meta-analysis. Oncotarget. 2017 Dec 23;9(4):5387-5396.
  • Rubio-Tapia A, Hill ID, Kelly CP, Calderwood AH, Murray JA; American College of Gastroenterology. ACG clinical guidelines: diagnosis and management of celiac disease. Am J Gastroenterol. 2013 May;108(5):656-76; quiz 677.
  • Volta U, Caio G, Tovoli F, De Giorgio R. Non-celiac gluten sensitivity: questions still to be answered despite increasing awareness. Cell Mol Immunol. 2013 Sep;10(5):383-92. doi: 10.1038/cmi.2013.28. Epub 2013 Aug 10. PMID: 23934026; PMCID: PMC4003198.
  • Lacy, Brian E. PhD, MD, FACG1; Pimentel, Mark MD, FACG2; Brenner, Darren M. MD, FACG3; Chey, William D. MD, FACG4; Keefer, Laurie A. PhD5; Long, Millie D. MDMPH, FACG (GRADE Methodologist)6; Moshiree, Baha MD, MSc, FACG7. ACG Clinical Guideline: Management of Irritable Bowel Syndrome. The American Journal of Gastroenterology 116(1):p 17-44, January 2021.