Published: July 2026
Medically reviewed by:
Gary Falcetano, PA-C, AE-C
Weed pollens are among the leading causes of seasonal allergic rhinitis, a condition estimated to affect up to 20% of adults worldwide.¹ Yet despite its prevalence, diagnosing the cause of seasonal allergy symptoms in primary care is not always straightforward.
Symptoms such as sneezing, nasal congestion, itchy eyes, and cough can overlap with other respiratory conditions, making it difficult to identify the underlying trigger based on symptoms alone. Clinical history is an important part of the patient's story. ImmunoCAPTM specific IgE testing can help identify allergen sensitizations and support diagnosis, management, and referral decisions.2
Start with clinical history
Clinical history is the foundation for evaluating patients with suspected weed pollen allergy. Symptom timing, seasonality, and environmental exposures may help determine whether pollen exposure contributes to a patient's symptoms.2
When evaluating patients with suspected weed pollen allergy, consider:
- Symptom timing and seasonality, particularly symptoms that recur during late summer or early fall
- Environmental exposures, including time spent outdoors, occupational exposures, and local pollen patterns
- Geographic location, as weed pollen prevalence and seasonality vary by region
- Respiratory symptoms, including wheeze or worsening asthma symptoms during pollen season
- Associated food reactions, such as itching or tingling of the lips, mouth, or throat after eating certain fruits or vegetables that may be associated with pollen-food allergy syndrome2
Clinical history guides the diagnostic evaluation, but it may not always identify the underlying trigger.2 ImmunoCAP Specific IgE testing can provide additional information about allergen sensitization to support clinical decision-making.
Recognizing weed pollen allergy
Weed pollens, including common ragweed, mugwort, plantain, and wall pellitory, are common causes of seasonal allergic rhinitis and may also contribute to allergic asthma symptoms.2 The most clinically relevant weed pollens vary by geographic region and pollen season.
Patients with weed pollen allergy may present with:2
- Sneezing
- Nasal congestion
- Rhinorrhea
- Itchy or watery eyes
- Cough
- Wheeze
- Worsening asthma symptoms during pollen season
These symptoms are not specific to allergy and may overlap with:2
- Viral upper respiratory infections
- Nonallergic rhinitis
- Chronic rhinosinusitis
- Asthma triggered by other environmental exposures
Because symptoms alone may not identify the underlying trigger, objective testing for allergen sensitization can help determine whether weed pollen sensitization may be contributing to a patient's symptoms. Together with clinical history, this provides a more complete picture to support diagnosis and patient management.
How specific IgE blood testing supports the diagnosis of weed pollen allergy
ImmunoCAP Specific IgE tests provide objective information about allergen sensitization to support clinical decision-making.
Using a routine blood sample, the tests help identify sensitization to clinically relevant weed pollen allergens. When interpreted alongside clinical history, symptoms and seasonal exposure patterns, results support a more informed diagnostic evaluation and patient management.
When identifying the trigger improves patient care
When weed pollen is confirmed as a clinically relevant trigger, primary care providers can move beyond symptom-based management to a more targeted approach to care. Identifying allergen sensitization can help clarify the potential triggers of seasonal symptoms, support diagnostic confidence, and provide a stronger foundation for treatment planning.
How to order an ImmunoCAP Specific IgE test
ImmunoCAP Specific IgE tests can be ordered through national and local laboratories. To help streamline the ordering process, the Lab Ordering Guide (LOG) provides laboratory-specific test information, searchable results by ZIP code and allergy profile type, and easy access to the codes needed for ordering.
Frequently asked questions
A detailed medical history is the first step. Evaluating symptom timing, seasonality, and environmental exposures can help determine whether weed pollen may be contributing to a patient's symptoms. Clinical history helps guide diagnostic evaluation and decisions about further testing.
When a patient's symptoms and history are consistent with weed pollen allergy, ImmunoCAP Specific IgE tests can help build a more complete picture by identifying allergen sensitizations and adding objective information to the clinical evaluation.
No. A positive specific IgE result indicates sensitization to an allergen, but sensitization alone does not confirm clinical allergy. Results should be interpreted alongside the patient's symptoms, clinical history, and exposure patterns to determine whether the sensitization is contributing to the patient's symptoms.2
Yes. In sensitized individuals, exposure to weed pollen can trigger or worsen allergic asthma symptoms, particularly during peak pollen season. Identifying pollen sensitizations may help explain seasonal respiratory symptoms, support differentiation of allergic and non-allergic causes of symptoms and inform diagnosis and management decisions.2