Talk with Us
Grass pollen allergy: What primary care providers need to know
Article
Allergic asthma
Respiratory allergy
Published: May 2026
Medically reviewed by: 
Gary Falcetano, PA-C, AE-C
Grass pollen season arrives every year, and so do the same patients presenting with all-too-familiar symptoms like congestion, sneezing, and itchy eyes. Treatment is started, adjusted, and often repeated, but many return still symptomatic and without a clear understanding of what is driving their condition.
For primary care providers, this pattern is common. But it raises an important question:
Are we managing symptoms, or identifying the triggers?
Grass pollen allergies are often assumed based on seasonality. However, overlapping symptoms, multiple sensitizations, and variable exposure make the underlying trigger not always clear.1  Without diagnostic clarity, care can become reactive, leading to unnecessary repeat visits and ongoing patient frustration.
Specific IgE testing in primary care provides an opportunity to change that pattern. Helping to determine or rule out grass pollen sensitization provides objective insight and supports more targeted, effective patient management.1
Brown eggs and a glass of cow's milk.
Understanding grass pollen exposure and patterns
The grass pollen season varies by region, with peak periods typically occurring in late spring to early summer.2  Timing and intensity can differ depending on local geography, climate, and grass species.
Grass pollen exposure is often characterized by short periods of high pollen counts. During these peaks, patients may experience a sudden onset or worsening of symptoms, even if they were previously mild.2 This can make symptom progression feel unpredictable.
Grass pollen levels are also influenced by environmental conditions:2
  • Higher on warm, dry, and windy days
  • Lower during rain or high humidity
These changes in pollen levels directly affect patient exposure. Symptoms often worsen with outdoor exposure on high-pollen days and improve when exposure is reduced.
Grass pollen season may overlap with other pollen seasons, such as tree pollen earlier in the year or weed pollen later, making it more difficult to attribute symptoms to a single trigger based on timing alone.1
Why are grass pollen symptoms tricky?
Symptoms don’t always point to a single cause
Grass pollen allergy symptoms often overlap with viral infections, nonallergic rhinitis, and sensitizations to other allergens.2 This can make it difficult to identify grass pollen as the main driver of symptoms based on presentation alone.
Exposure and symptoms can be inconsistent
Grass pollen allergy can present with a combination of nasal and ocular symptoms and may also be associated with lower airway symptoms such as cough, wheeze, or shortness of breath, reflecting the link between allergic rhinitis and asthma.3
Day-to-day changes in pollen levels, environmental conditions, and exposure can cause symptoms to vary, making patterns harder to interpret.
Multiple allergens are often the triggers
Some patients are sensitized to multiple allergens. Overlapping pollen seasons or additional triggers can extend symptom duration, making it difficult to identify a single trigger based on the time of year.1
Specific IgE blood testing for grass pollen sensitization
Given these diagnostic challenges, identifying the underlying trigger is important.
With one blood draw, ImmunoCAP™ Specific IgE tests can help determine whether grass pollen may be contributing to a patient’s symptoms or if another trigger is involved. This is especially helpful when symptoms:
  • Last longer than expected
  • Do not respond to treatment
  • Appear inconsistent or unclear
Testing can identify sensitizations to other allergens, such as tree pollen or weeds, which may explain why symptoms persist across the season.
Learn more about ImmunoCAP Specific IgE testing and its role in allergy diagnosis.
The role of specific IgE blood testing in primary care
When you know grass pollen is the trigger, management becomes more straightforward.
Instead of adjusting treatment based solely on symptoms, you can align your approach with what drives them. This may include timing treatment to pollen peaks, setting expectations, or recognizing when symptoms are related to exposure rather than treatment failure. Identifying the triggers earlier can help to reduce unnecessary repeat visits and support more effective symptom control.
It also supports clearer conversations with patients. When patients understand how pollen exposure affects their symptoms, they are better able to anticipate and manage them.
How to order an ImmunoCAP Specific IgE test
ImmunoCAP Specific IgE blood tests can be ordered through national or local laboratories. The Lab Ordering Guide (LOG) helps you find the right allergy profile test codes with confidence—allowing you to filter by ZIP code and allergy profile type, access customized results from labs you already use, and easily copy codes for ordering.
Access the Lab Ordering Guide to streamline ordering and select the most relevant tests for your patients.
Grass pollen allergy FAQs
If symptoms persist despite treatment, consider reassessing the diagnosis, adherence, and trigger exposure. Confirm that the patient is using medications correctly and consistently, and review ongoing allergen exposure, including seasonal and environmental factors. Specific IgE testing can help identify allergen sensitization when the underlying trigger remains unclear, supporting more informed clinical decision-making. Step up therapy as appropriate or consider referral to an allergy specialist for further evaluation.
Order ImmunoCAP Specific IgE tests for grass pollen when patients present with persistent or seasonal rhinitis or conjunctivitis symptoms that suggest an allergic cause. Testing is appropriate when symptoms are moderate to severe, not well controlled with empiric treatment, or when there is suspicion of sensitization to multiple allergens. It may be particularly useful when symptoms are prolonged, inconsistent, or difficult to attribute to a single trigger based on seasonality alone. Identifying the relevant allergen(s) can help guide management decisions, such as targeted avoidance strategies or referral for specialist care.
Managing grass pollen allergy effectively combines allergen avoidance, medication as needed, and patient education. Advise patients to minimize exposure during peak grass pollen seasons and to optimize first-line therapies, such as intranasal corticosteroids and non-sedating antihistamines. If symptoms remain uncontrolled, consider referral to a specialist for further evaluation. 
Primary care providers can guide patients to reduce grass pollen exposure with simple, practical steps. Advise patients to check daily pollen counts and limit outdoor activities during peak times, keep windows closed during pollen season, and use air conditioning when possible. After being outdoors, patients should shower, wash their hair, and change clothes to remove pollen. Drying laundry indoors and wearing sunglasses outdoors may also help reduce exposure. These small actions can help improve symptom control and quality of life.
Refer patients to an allergy specialist if symptoms persist despite treatment or the diagnosis is unclear. Referral is also advised when considering allergen immunotherapy, since specialist assessment is necessary to evaluate appropriateness and guide therapy. Patients with severe or difficult-to-control asthma, especially if an allergic trigger is suspected, may benefit from specialist care. Additionally, refer patients with sensitization to multiple allergens, as further detailed evaluation can identify relevant triggers and enable more targeted management.
References
  1. Wise SK, et al. International Consensus Statement on Allergy and Rhinology: Allergic Rhinitis. Int Forum Allergy Rhinol. 2023;13:293–859.
  2. D’Amato G, et al. Allergenic pollen and pollen allergy in Europe. Allergy. 2016;71:154–165.
  3. Brożek JL, et al. Allergic Rhinitis and its Impact on Asthma (ARIA) guidelines—2020 revision. J Allergy Clin Immunol. 2020;145:70–80.