Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp6nl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080548.1 | NP_001074017.1 | 10 - 11 | 950 | 138 | |
NM_181399.3 | NP_852064.2 | 9 - 10 | 789 | 138 | ||
GenBank mRNA | AK084045.1 | - | 10 - 11 | 774 | 138 | |
AK129037.1 | - | 10 - 11 | 802 | 138 | ||
AK155073.1 | - | 10 - 11 | 951 | 138 | ||
BC051184.1 | - | 9 - 10 | 792 | 138 | ||
CN455065.1 | - | 2 - 3 | 179 | 138 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Mus musculus Probe spans exons |