The Thermo Fisher Scientific Accula system is a rapid diagnostic point of care platform that provides simple, accurate sample-to-answer rapid PCR.
The Accula System consists of the Accula Dock and Accula Test Kit. The dock is a small, reusable device about the size of your palm that holds a cassette. It plugs into a standard electrical outlet and provides power to the cassette, which contains the components and reagents for viral lysis, reverse transcription, and PCR amplification. The Accula rapid PCR Tests provide reliable, qualitative PCR results in ~30 minutes.
Accula rapid PCR testing products
Accula Dock
The Accula Dock consists of an electro-mechanical interface to a single test cassette with all electrical systems, controls, and logic necessary to orchestrate an in vitro diagnostic test.
Accula SARS-CoV-2 Test
The Accula SARS-CoV-2 Test combines the accuracy of PCR with the speed and simplicity of rapid antigen tests, delivering visual results in ~30 minutes.
Workflow
The Accula System workflow is elegantly simple: swab, elute, and follow the prompts on the dock to load the sample and run the test. Hands-on time is ~1 minute, after which you can walk away. The dock notifies you to read the results when the test is complete.
How it works
The Accula System is a rapid diagnostic platform that combines the accuracy of RT-PCR with the simplicity, convenience, and procedural familiarity of traditional rapid immunoassays. The Accula SARS-CoV-2 Test has received an Emergency Use Authorization (EUA) from the FDA for the detection of SARS-CoV-2 in Clinical Laboratory Improvement Amendments (CLIA)–waived environments. Designed for use with the palm-sized Accula Dock, the Accula SARS-CoV-2 Test provides reliable, qualitative results in approximately 30 minutes.
What customers are saying
Featured articles
Available through these distributors
Contact us
Technical support
Email: techsupport@thermofisher.com
Phone: (800) 955 6288, option 2
Customer support
Email: USSD2.customercare@thermofisher.com
Phone: (858) 800-4929
For Emergency Use Authorization (EUA) Only. For prescription use only. For in vitro diagnostic use.