Chemically Defined Lipid Concentrate is a concentrated lipid emulsion designed to reduce or replace fetal bovine serum in cell culture media for a wide variety of applications, including growth and maintenance of CHO, hybridoma, and insect cells in culture; monoclonal antibody production by hybridomas; and viral expression in insect cells. The components of this medium supplement are chemically-defined.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell Type
Mammalian Cells
Form
Liquid
Product Type
Lipid Concentrate
Shelf Life
12 Months
Green Features
Sustainable packaging
pH
4 to 6
Shipping Condition
Room Temperature
Contents & Storage
Store in refrigerator (2–8°C) and protect from light.