Product References
Tyrosine Kinase Inhibitors Target B Lymphocytes.
Biomolecules
Upfold NLE,Petakh P,Kamyshnyi A,Oksenych V
Published figure using CD19 monoclonal antibody (Product # 12-0198-42) in Flow Cytometry
Sat Feb 25 00:00:00 EST 2023
Evaluation of peripheral lymphocyte subsets in acne vulgaris patients before and after systemic isotretinoin treatment.
Indian journal of pharmacology
Gunaydin SD,Tezcan I
Published figure using CD19 monoclonal antibody (Product # 12-0198-42) in Flow Cytometry
Wed Dec 21 00:00:00 EST 2022
Diurnal Variation of Plasma Extracellular Vesicle Is Disrupted in People Living with HIV.
Pathogens (Basel, Switzerland)
Bazié WW,Goyer B,Boucher J,Zhang Y,Planas D,Chatterjee D,Routy JP,Alary M,Ancuta P,Gilbert C
12-0198-42 was used in Flow Cytometry to investigate daily variations of large and small EVs, in terms of abundance and microRNA contents in people living with HIV (PLWH) receiving antiretroviral therapy (HIV+ART) and uninfected controls (HIV-).
Sat Apr 24 00:00:00 EDT 2021
Innate-like self-reactive B cells infiltrate human renal allografts during transplant rejection.
Nature communications
Asano Y,Daccache J,Jain D,Ko K,Kinloch A,Veselits M,Wolfgeher D,Chang A,Josephson M,Cunningham P,Tambur A,Khan AA,Pillai S,Chong AS,Clark MR
12-0198-42 was used in Flow Cytometry to show a mechanism of human inflammation in which a breach in organ-restricted tolerance by infiltrating innate-like B cells drives local tissue destruction.
Fri Jul 16 00:00:00 EDT 2021
Innate-like self-reactive B cells infiltrate human renal allografts during transplant rejection.
Nature communications
Asano Y,Daccache J,Jain D,Ko K,Kinloch A,Veselits M,Wolfgeher D,Chang A,Josephson M,Cunningham P,Tambur A,Khan AA,Pillai S,Chong AS,Clark MR
12-0198-42 was used in Flow Cytometry to show a mechanism of human inflammation in which a breach in organ-restricted tolerance by infiltrating innate-like B cells drives local tissue destruction.
Fri Jul 16 00:00:00 EDT 2021
Phase I Study of Ficlatuzumab and Cetuximab in Cetuximab-Resistant, Recurrent/Metastatic Head and Neck Cancer.
Cancers
Bauman JE,Ohr J,Gooding WE,Ferris RL,Duvvuri U,Kim S,Johnson JT,Soloff AC,Wallweber G,Winslow J,Gaither-Davis A,Grandis JR,Stabile LP
12-0198-42 was used in Flow Cytometry to establish the recommended Phase II dose (RP2D). Secondary objectives included overall response rate (ORR), progression-free survival (PFS), and overall survival (OS).
Thu Jun 11 00:00:00 EDT 2020
The downregulated membrane expression of CD18 in CD34+ cells defines a primitive population of human hematopoietic stem cells.
Stem cell research & therapy
Mesa-Núñez C,Leon-Rico D,Aldea M,Damián C,Sanchez-Baltasar R,Sanchez R,Alberquilla O,Segovia JC,Bueren JA,Almarza E
Published figure using CD19 monoclonal antibody (Product # 12-0198-42) in Flow Cytometry
Tue Apr 28 00:00:00 EDT 2020
Human C. difficile toxin-specific memory B cell repertoires encode poorly neutralizing antibodies.
JCI insight
Shah HB,Smith K,Scott EJ,Larabee JL,James JA,Ballard JD,Lang ML
Published figure using CD19 monoclonal antibody (Product # 12-0198-42) in Flow Cytometry
Thu Aug 20 00:00:00 EDT 2020
Umbilical cord blood‑derived Helios‑positive regulatory T cells promote angiogenesis in acute lymphoblastic leukemia in mice via CCL22 and the VEGFA‑VEGFR2 pathway.
Molecular medicine reports
Li X,Li D,Shi Q,Huang X,Ju X
12-0198 was used in Flow cytometry/Cell sorting to elucidate the mechanisms underlying the role of regulatory T cells in acute lymphoblastic leukaemia.
Wed May 01 00:00:00 EDT 2019
Evaluation of Canonical Inflammasome Activation in Human Monocytes by Imaging Flow Cytometry.
Frontiers in immunology
Lage SL,Dominical VM,Wong CS,Sereti I
12-0198-42 was used in Flow Cytometry to explore the optimal imaging-based tools to measure ASC (apoptosis-associated speck-like protein containing a caspase activating and recruitment domain) speck formation.
Wed Oct 28 00:00:00 EDT 2020
Multi-Donor Longitudinal Antibody Repertoire Sequencing Reveals the Existence of Public Antibody Clonotypes in HIV-1 Infection.
Cell host & microbe
Setliff I,McDonnell WJ,Raju N,Bombardi RG,Murji AA,Scheepers C,Ziki R,Mynhardt C,Shepherd BE,Mamchak AA,Garrett N,Karim SA,Mallal SA,Crowe JE,Morris L,Georgiev IS
12-0198 was used in Flow cytometry/Cell sorting to reveal the existence of "public" antibody clonotypes that were shared among multiple HIV-infected individuals.
Wed Jun 13 00:00:00 EDT 2018
Bacillus anthracis lethal toxin negatively modulates ILC3 function through perturbation of IL-23-mediated MAPK signaling.
PLoS pathogens
Seshadri S,Allan DSJ,Carlyle JR,Zenewicz LA
12-0198 was used in Flow cytometry/Cell sorting to study the effects of MAPK signalling pathways in group 3 innate immune lymphocytes using Bacillus anthracis.
Sun Oct 01 00:00:00 EDT 2017
The Influence of Cytomegalovirus on Expression of HLA-G and its Ligand KIR2DL4 by Human Peripheral Blood Leucocyte Subsets.
Scandinavian journal of immunology
Albayati Z,Alyami A,Alomar S,Middleton D,Bonnett L,Aleem S,Flanagan BF,Christmas SE
Published figure using CD19 monoclonal antibody (Product # 12-0198-42) in Flow Cytometry
Wed Nov 01 00:00:00 EDT 2017
Effects of acute exposure to low-dose radiation on the characteristics of human bone marrow mesenchymal stromal/stem cells.
Inflammation and regeneration
Fujishiro A,Miura Y,Iwasa M,Fujii S,Sugino N,Andoh A,Hirai H,Maekawa T,Ichinohe T
12-0198 was used in Flow cytometry/Cell sorting to determine whether low dose irradiation affects the function of mesenchymal stromal/stem cells.
Fri Mar 11 00:00:00 EST 2022
Long-lived Plasmodium falciparum specific memory B cells in naturally exposed Swedish travelers.
European journal of immunology
Ndungu FM,Lundblom K,Rono J,Illingworth J,Eriksson S,Färnert A
Published figure using CD19 monoclonal antibody (Product # 12-0198-42) in Flow Cytometry
Fri Nov 01 00:00:00 EDT 2013
Chronic exposure to Plasmodium falciparum is associated with phenotypic evidence of B and T cell exhaustion.
Journal of immunology (Baltimore, Md. : 1950)
Illingworth J,Butler NS,Roetynck S,Mwacharo J,Pierce SK,Bejon P,Crompton PD,Marsh K,Ndungu FM
Published figure using CD19 monoclonal antibody (Product # 12-0198-42) in Flow Cytometry
Fri Feb 01 00:00:00 EST 2013