Product References
APOLD1 loss causes endothelial dysfunction involving cell junctions, cytoskeletal architecture, and Weibel-Palade bodies, while disrupting hemostasis.
Haematologica
Stritt S,Nurden P,Nurden AT,Schved JF,Bordet JC,Roux M,Alessi MC,Trégouët DA,Mäkinen T,Giansily-Blaizot M
14-0411-82 was used in Immunocytochemistry-immunoflourescence to identify APOLD1 as an important regulator of vascular homeostasis and raise the need to consider testing of EC function in patients with IBDs without apparent platelet or coagulation defects.
Wed Mar 01 00:00:00 EST 2023
SNAP23 is essential for platelet and mast cell development and required in connective tissue mast cells for anaphylaxis.
The Journal of biological chemistry
Cardenas RA,Gonzalez R,Sanchez E,Ramos MA,Cardenas EI,Rodarte AI,Alcazar-Felix RJ,Isaza A,Burns AR,Heidelberger R,Adachi R
Published figure using CD41a monoclonal antibody (Product # 14-0411-82) in Flow Cytometry
Mon Aug 02 00:00:00 EDT 2021
All-trans-retinoic acid shifts Th1 towards Th2 cell differentiation by targeting NFAT1 signalling to ameliorate immune-mediated aplastic anaemia.
British journal of haematology
Tang D,Liu S,Sun H,Qin X,Zhou N,Zheng W,Zhang M,Zhou H,Tuersunayi A,Duan C,Chen J
Published figure using CD41a monoclonal antibody (Product # 14-0411-82) in Immunohistochemistry
Tue Dec 01 00:00:00 EST 2020
Single-nucleotide human disease mutation inactivates a blood-regenerative GATA2 enhancer.
The Journal of clinical investigation
Soukup AA,Zheng Y,Mehta C,Wu J,Liu P,Cao M,Hofmann I,Zhou Y,Zhang J,Johnson KD,Choi K,Keles S,Bresnick EH
Published figure using CD41a monoclonal antibody (Product # 14-0411-82) in Flow Cytometry
Fri Mar 01 00:00:00 EST 2019
Iron deficiency disrupts embryonic haematopoiesis but not the endothelial to haematopoietic transition.
Scientific reports
Shvartsman M,Bilican S,Lancrin C
Published figure using CD41a monoclonal antibody (Product # 14-0411-82) in Flow Cytometry
Tue Apr 23 00:00:00 EDT 2019
The role of platelets in mediating a response to human influenza infection.
Nature communications
Koupenova M,Corkrey HA,Vitseva O,Manni G,Pang CJ,Clancy L,Yao C,Rade J,Levy D,Wang JP,Finberg RW,Kurt-Jones EA,Freedman JE
Published figure using CD41a monoclonal antibody (Product # 14-0411-82) in Immunocytochemistry
Tue Apr 16 00:00:00 EDT 2019
Adrenergic nerve degeneration in bone marrow drives aging of the hematopoietic stem cell niche.
Nature medicine
Maryanovich M,Zahalka AH,Pierce H,Pinho S,Nakahara F,Asada N,Wei Q,Wang X,Ciero P,Xu J,Leftin A,Frenette PS
Published figure using CD41a monoclonal antibody (Product # 14-0411-82) in Immunofluorescence
Fri Jun 01 00:00:00 EDT 2018
JAK2/IDH-mutant-driven myeloproliferative neoplasm is sensitive to combined targeted inhibition.
The Journal of clinical investigation
McKenney AS,Lau AN,Somasundara AVH,Spitzer B,Intlekofer AM,Ahn J,Shank K,Rapaport FT,Patel MA,Papalexi E,Shih AH,Chiu A,Freinkman E,Akbay EA,Steadman M,Nagaraja R,Yen K,Teruya-Feldstein J,Wong KK,Rampal R,Vander Heiden MG,Thompson CB,Levine RL
14-0411 was used in Flow cytometry/Cell sorting to suggest that combined JAK and IDH inhibition may offer a therapeutic advantage in this high-risk MPN subtype.
Thu Feb 01 00:00:00 EST 2018
Transient loss of venous integrity during developmental vascular remodeling leads to red blood cell extravasation and clearance by lymphatic vessels.
Development (Cambridge, England)
Zhang Y,Daubel N,Stritt S,Mäkinen T
Published figure using CD41a monoclonal antibody (Product # 14-0411-82) in Immunofluorescence
Thu Feb 08 00:00:00 EST 2018
Single-cell transcriptomics reveals a new dynamical function of transcription factors during embryonic hematopoiesis.
eLife
Bergiers I,Andrews T,Vargel Bölükbaşı Ö,Buness A,Janosz E,Lopez-Anguita N,Ganter K,Kosim K,Celen C,Itır Perçin G,Collier P,Baying B,Benes V,Hemberg M,Lancrin C
Published figure using CD41a monoclonal antibody (Product # 14-0411-82) in Flow Cytometry
Tue Mar 20 00:00:00 EDT 2018
Single-cell transcriptomics reveals a new dynamical function of transcription factors during embryonic hematopoiesis.
eLife
Bergiers I,Andrews T,Vargel Bölükbaşı Ö,Buness A,Janosz E,Lopez-Anguita N,Ganter K,Kosim K,Celen C,Itır Perçin G,Collier P,Baying B,Benes V,Hemberg M,Lancrin C
Published figure using CD41a monoclonal antibody (Product # 14-0411-82) in Flow Cytometry
Tue Mar 20 00:00:00 EDT 2018
Cathelicidins prime platelets to mediate arterial thrombosis and tissue inflammation.
Nature communications
Pircher J,Czermak T,Ehrlich A,Eberle C,Gaitzsch E,Margraf A,Grommes J,Saha P,Titova A,Ishikawa-Ankerhold H,Stark K,Petzold T,Stocker T,Weckbach LT,Novotny J,Sperandio M,Nieswandt B,Smith A,Mannell H,Walzog B,Horst D,Soehnlein O,Massberg S,Schulz C
14-0411 was used in Immunohistochemistry-immunofluorescence to understand the role of cathelicidin LL-37 in thrombosis and tissue inflammation.
Wed Apr 18 00:00:00 EDT 2018
Cathelicidins prime platelets to mediate arterial thrombosis and tissue inflammation.
Nature communications
Pircher J,Czermak T,Ehrlich A,Eberle C,Gaitzsch E,Margraf A,Grommes J,Saha P,Titova A,Ishikawa-Ankerhold H,Stark K,Petzold T,Stocker T,Weckbach LT,Novotny J,Sperandio M,Nieswandt B,Smith A,Mannell H,Walzog B,Horst D,Soehnlein O,Massberg S,Schulz C
14-0411 was used in Immunohistochemistry-immunofluorescence to understand the role of cathelicidin LL-37 in thrombosis and tissue inflammation.
Wed Apr 18 00:00:00 EDT 2018
Differential cytokine contributions of perivascular haematopoietic stem cell niches.
Nature cell biology
Asada N,Kunisaki Y,Pierce H,Wang Z,Fernandez NF,Birbrair A,Ma'ayan A,Frenette PS
Published figure using CD41a monoclonal antibody (Product # 14-0411-82) in Immunofluorescence
Wed Mar 01 00:00:00 EST 2017
A population of hematopoietic stem cells derives from GATA4-expressing progenitors located in the placenta and lateral mesoderm of mice.
Haematologica
Cañete A,Carmona R,Ariza L,Sánchez MJ,Rojas A,Muñoz-Chápuli R
14041182 was used in flow cytometry to utilize a G2-Gata4Cre;R26REYFP mouse line to track the developmental fate of the G2-Gata4 cell lineage
Sat Apr 01 00:00:00 EDT 2017
Assessment of Th17/Treg cells and Th cytokines in an improved immune thrombocytopenia mouse model.
Hematology (Amsterdam, Netherlands)
Zhang G,Zhang P,Liu H,Liu X,Xie S,Wang X,Wu Y,Chang J,Ma L
14-0411-82 was used in Miscellaneous to suggest that the improved passive ITP mouse model does not mimic the autoimmune inflammatory process of human ITP.
Fri Sep 01 00:00:00 EDT 2017
Mediator Kinase Phosphorylation of STAT1 S727 Promotes Growth of Neoplasms With JAK-STAT Activation.
EBioMedicine
Nitulescu II,Meyer SC,Wen QJ,Crispino JD,Lemieux ME,Levine RL,Pelish HE,Shair MD
Published figure using CD41a monoclonal antibody (Product # 14-0411-82) in Flow Cytometry
Fri Dec 01 00:00:00 EST 2017
Targeted genome editing restores T cell differentiation in a humanized X-SCID pluripotent stem cell disease model.
Scientific reports
Alzubi J,Pallant C,Mussolino C,Howe SJ,Thrasher AJ,Cathomen T
Published figure using CD41a monoclonal antibody (Product # 14-0411-82) in Flow Cytometry
Fri Sep 29 00:00:00 EDT 2017
Leukaemogenic effects of Ptpn11 activating mutations in the stem cell microenvironment.
Nature
Dong L,Yu WM,Zheng H,Loh ML,Bunting ST,Pauly M,Huang G,Zhou M,Broxmeyer HE,Scadden DT,Qu CK
14041182 was used in immunohistochemistry - frozen section to elucidate the contribution of Ptpn11 mutations in the bone marrow microenvironment to leukaemogenesis
Thu Nov 10 00:00:00 EST 2016
Hematopoietic Stem Cells Count and Remember Self-Renewal Divisions.
Cell
Bernitz JM,Kim HS,MacArthur B,Sieburg H,Moore K
Published figure using CD41a monoclonal antibody (Product # 14-0411-82) in Flow Cytometry
Thu Nov 17 00:00:00 EST 2016
Dicer1-mediated miRNA processing shapes the mRNA profile and function of murine platelets.
Blood
Rowley JW,Chappaz S,Corduan A,Chong MM,Campbell R,Khoury A,Manne BK,Wurtzel JG,Michael JV,Goldfinger LE,Mumaw MM,Nieman MT,Kile BT,Provost P,Weyrich AS
14-0411 was used in Flow cytometry/Cell sorting to study the role of Dicer1 in the regulation of integrin expression by platelets.
Thu Apr 07 00:00:00 EDT 2016
Transcriptome-based profiling of yolk sac-derived macrophages reveals a role for Irf8 in macrophage maturation.
The EMBO journal
Hagemeyer N,Kierdorf K,Frenzel K,Xue J,Ringelhan M,Abdullah Z,Godin I,Wieghofer P,Costa Jordão MJ,Ulas T,Yorgancioglu G,Rosenbauer F,Knolle PA,Heikenwalder M,Schultze JL,Prinz M
14-0411 was used in Immunohistochemistry-wholemount to study the cellular and molecular mechanisms which govern the origin and developmental pathways of tissue macrophages.
Mon Aug 15 00:00:00 EDT 2016
The Poly(C) Binding Protein Pcbp2 and Its Retrotransposed Derivative Pcbp1 Are Independently Essential to Mouse Development.
Molecular and cellular biology
Ghanem LR,Kromer A,Silverman IM,Chatterji P,Traxler E,Penzo-Mendez A,Weiss MJ,Stanger BZ,Liebhaber SA
Published figure using CD41a monoclonal antibody (Product # 14-0411-82) in Flow Cytometry
Fri Jan 15 00:00:00 EST 2016
Murine iPSC-Derived Macrophages as a Tool for Disease Modeling of Hereditary Pulmonary Alveolar Proteinosis due to Csf2rb Deficiency.
Stem cell reports
Mucci A,Kunkiel J,Suzuki T,Brennig S,Glage S,Kühnel MP,Ackermann M,Happle C,Kuhn A,Schambach A,Trapnell BC,Hansen G,Moritz T,Lachmann N
Published figure using CD41a monoclonal antibody (Product # 14-0411-82) in Flow Cytometry
Tue Aug 09 00:00:00 EDT 2016
Activation of the TGFβ pathway impairs endothelial to haematopoietic transition.
Scientific reports
Vargel Ö,Zhang Y,Kosim K,Ganter K,Foehr S,Mardenborough Y,Shvartsman M,Enright AJ,Krijgsveld J,Lancrin C
Published figure using CD41a monoclonal antibody (Product # 14-0411-82) in Flow Cytometry
Fri Feb 19 00:00:00 EST 2016
Small Molecule Inhibitor of CBFβ-RUNX Binding for RUNX Transcription Factor Driven Cancers.
EBioMedicine
Illendula A,Gilmour J,Grembecka J,Tirumala VSS,Boulton A,Kuntimaddi A,Schmidt C,Wang L,Pulikkan JA,Zong H,Parlak M,Kuscu C,Pickin A,Zhou Y,Gao Y,Mishra L,Adli M,Castilla LH,Rajewski RA,Janes KA,Guzman ML,Bonifer C,Bushweller JH
Published figure using CD41a monoclonal antibody (Product # 14-0411-82) in Flow Cytometry
Wed Jun 01 00:00:00 EDT 2016
Novel function of FAXDC2 in megakaryopoiesis.
Blood cancer journal
Jin Q,Ren Y,Wang M,Suraneni PK,Li D,Crispino JD,Fan J,Huang Z
Published figure using CD41a monoclonal antibody (Product # 14-0411-82) in Flow Cytometry
Fri Sep 30 00:00:00 EDT 2016
EphrinB2 regulates the emergence of a hemogenic endothelium from the aorta.
Scientific reports
Chen II,Caprioli A,Ohnuki H,Kwak H,Porcher C,Tosato G
Published figure using CD41a monoclonal antibody (Product # 14-0411-82) in Immunocytochemistry
Thu Jun 02 00:00:00 EDT 2016
EphrinB2 regulates the emergence of a hemogenic endothelium from the aorta.
Scientific reports
Chen II,Caprioli A,Ohnuki H,Kwak H,Porcher C,Tosato G
Published figure using CD41a monoclonal antibody (Product # 14-0411-82) in Immunocytochemistry
Thu Jun 02 00:00:00 EDT 2016
H2S protects against fatal myelosuppression by promoting the generation of megakaryocytes/platelets.
Journal of hematology & oncology
Liu HD,Zhang AJ,Xu JJ,Chen Y,Zhu YC
Published figure using CD41a monoclonal antibody (Product # 14-0411-82) in Flow Cytometry
Wed Feb 24 00:00:00 EST 2016
H2S protects against fatal myelosuppression by promoting the generation of megakaryocytes/platelets.
Journal of hematology & oncology
Liu HD,Zhang AJ,Xu JJ,Chen Y,Zhu YC
Published figure using CD41a monoclonal antibody (Product # 14-0411-82) in Flow Cytometry
Wed Feb 24 00:00:00 EST 2016
Tumor Suppressor Analysis in CML.
Methods in molecular biology (Clifton, N.J.)
Herrmann O,Schemionek M
14041182 was used in flow cytometry to describe a method for the retroviral insertion of Bcr-Abl into mice that are genetically depleted for a potential tumor suppressor
Fri Jan 26 00:00:00 EST 2018
Role of TP53 mutations in the origin and evolution of therapy-related acute myeloid leukaemia.
Nature
Wong TN,Ramsingh G,Young AL,Miller CA,Touma W,Welch JS,Lamprecht TL,Shen D,Hundal J,Fulton RS,Heath S,Baty JD,Klco JM,Ding L,Mardis ER,Westervelt P,DiPersio JF,Walter MJ,Graubert TA,Ley TJ,Druley T,Link DC,Wilson RK
14-0411 was used in Flow cytometry/Cell sorting to investigate how prior exposure to cytotoxic therapy influences leukemogenesis and TP53 mutations in t-AML/t-MDS patients.
Thu Feb 26 00:00:00 EST 2015
Specific Inflammatory Stimuli Lead to Distinct Platelet Responses in Mice and Humans.
PloS one
Beaulieu LM,Clancy L,Tanriverdi K,Benjamin EJ,Kramer CD,Weinberg EO,He X,Mekasha S,Mick E,Ingalls RR,Genco CA,Freedman JE
Published figure using CD41a monoclonal antibody (Product # 14-0411-82) in Flow Cytometry
Fri Mar 25 00:00:00 EDT 2016
A Quantitative Proteomic Analysis of Hemogenic Endothelium Reveals Differential Regulation of Hematopoiesis by SOX17.
Stem cell reports
Clarke RL,Robitaille AM,Moon RT,Keller G
Published figure using CD41a monoclonal antibody (Product # 14-0411-82) in Flow Cytometry
Tue Aug 11 00:00:00 EDT 2015
Lysophosphatidic acid acts as a nutrient-derived developmental cue to regulate early hematopoiesis.
The EMBO journal
Li H,Yue R,Wei B,Gao G,Du J,Pei G
Published figure using CD41a monoclonal antibody (Product # 14-0411-82) in Flow Cytometry
Tue Jun 17 00:00:00 EDT 2014
Platelet glycoprotein Ib-IX as a regulator of systemic inflammation.
Arteriosclerosis, thrombosis, and vascular biology
Corken A,Russell S,Dent J,Post SR,Ware J
Published figure using CD41a monoclonal antibody (Product # 14-0411-82) in Flow Cytometry
Thu May 01 00:00:00 EDT 2014
Runx1 is required for progression of CD41+ embryonic precursors into HSCs but not prior to this.
Development (Cambridge, England)
Liakhovitskaia A,Rybtsov S,Smith T,Batsivari A,Rybtsova N,Rode C,de Bruijn M,Buchholz F,Gordon-Keylock S,Zhao S,Medvinsky A
Published figure using CD41a monoclonal antibody (Product # 14-0411-82) in Flow Cytometry
Mon Sep 01 00:00:00 EDT 2014
Runx1 is required for progression of CD41+ embryonic precursors into HSCs but not prior to this.
Development (Cambridge, England)
Liakhovitskaia A,Rybtsov S,Smith T,Batsivari A,Rybtsova N,Rode C,de Bruijn M,Buchholz F,Gordon-Keylock S,Zhao S,Medvinsky A
Published figure using CD41a monoclonal antibody (Product # 14-0411-82) in Flow Cytometry
Mon Sep 01 00:00:00 EDT 2014
Runx1 is required for progression of CD41+ embryonic precursors into HSCs but not prior to this.
Development (Cambridge, England)
Liakhovitskaia A,Rybtsov S,Smith T,Batsivari A,Rybtsova N,Rode C,de Bruijn M,Buchholz F,Gordon-Keylock S,Zhao S,Medvinsky A
Published figure using CD41a monoclonal antibody (Product # 14-0411-82) in Flow Cytometry
Mon Sep 01 00:00:00 EDT 2014
Single-cell analyses of regulatory network perturbations using enhancer-targeting TALEs suggest novel roles for PU.1 during haematopoietic specification.
Development (Cambridge, England)
Wilkinson AC,Kawata VK,Schütte J,Gao X,Antoniou S,Baumann C,Woodhouse S,Hannah R,Tanaka Y,Swiers G,Moignard V,Fisher J,Hidetoshi S,Tijssen MR,de Bruijn MF,Liu P,Göttgens B
Published figure using CD41a monoclonal antibody (Product # 14-0411-82) in Flow Cytometry
Wed Oct 01 00:00:00 EDT 2014
A crucial role for the ubiquitously expressed transcription factor Sp1 at early stages of hematopoietic specification.
Development (Cambridge, England)
Gilmour J,Assi SA,Jaegle U,Kulu D,van de Werken H,Clarke D,Westhead DR,Philipsen S,Bonifer C
Published figure using CD41a monoclonal antibody (Product # 14-0411-82) in Flow Cytometry
Sun Jun 01 00:00:00 EDT 2014
Expression of podocalyxin separates the hematopoietic and vascular potentials of mouse embryonic stem cell-derived mesoderm.
Stem cells (Dayton, Ohio)
Zhang H,Nieves JL,Fraser ST,Isern J,Douvaras P,Papatsenko D,D'Souza SL,Lemischka IR,Dyer MA,Baron MH
Published figure using CD41a monoclonal antibody (Product # 14-0411-82) in Flow Cytometry
Wed Jan 01 00:00:00 EST 2014
Mitochondrial anti-oxidant protects IEX-1 deficient mice from organ damage during endotoxemia.
International immunopharmacology
Ramsey H,Wu MX
14-0411 was used in Flow cytometry/Cell sorting to confirm significant contribution of mitochondrial ROS to the etiology of sepsis.
Mon Dec 01 00:00:00 EST 2014
PSTPIP2 dysregulation contributes to aberrant terminal differentiation in GATA-1-deficient megakaryocytes by activating LYN.
Cell death & disease
Liu L,Wen Q,Gong R,Gilles L,Stankiewicz MJ,Li W,Guo M,Li L,Sun X,Li W,Crispino JD,Huang Z
Published figure using CD41a monoclonal antibody (Product # 14-0411-82) in Flow Cytometry
Thu Jan 09 00:00:00 EST 2014
A directional switch of integrin signalling and a new anti-thrombotic strategy.
Nature
Shen B,Zhao X,O'Brien KA,Stojanovic-Terpo A,Delaney MK,Kim K,Cho J,Lam SC,Du X
Published figure using CD41a monoclonal antibody (Product # 14-0411-82) in Western Blot
Thu Nov 07 00:00:00 EST 2013
A directional switch of integrin signalling and a new anti-thrombotic strategy.
Nature
Shen B,Zhao X,O'Brien KA,Stojanovic-Terpo A,Delaney MK,Kim K,Cho J,Lam SC,Du X
Published figure using CD41a monoclonal antibody (Product # 14-0411-82) in Western Blot
Thu Nov 07 00:00:00 EST 2013
A directional switch of integrin signalling and a new anti-thrombotic strategy.
Nature
Shen B,Zhao X,O'Brien KA,Stojanovic-Terpo A,Delaney MK,Kim K,Cho J,Lam SC,Du X
Published figure using CD41a monoclonal antibody (Product # 14-0411-82) in Western Blot
Thu Nov 07 00:00:00 EST 2013
Podoplanin maintains high endothelial venule integrity by interacting with platelet CLEC-2.
Nature
Herzog BH,Fu J,Wilson SJ,Hess PR,Sen A,McDaniel JM,Pan Y,Sheng M,Yago T,Silasi-Mansat R,McGee S,May F,Nieswandt B,Morris AJ,Lupu F,Coughlin SR,McEver RP,Chen H,Kahn ML,Xia L
14-0411 was used in Immunohistochemistry to present a role for the transmembrane O-glycoprotein podoplanin (PDPN) in maintaining the barrier function of high endothelial venules.
Thu Oct 03 00:00:00 EDT 2013
Natural killer cells are required for extramedullary hematopoiesis following murine cytomegalovirus infection.
Cell host & microbe
Jordan S,Ruzsics Z,Mitrović M,Baranek T,Arapović J,Krmpotić A,Vivier E,Dalod M,Jonjić S,Dölken L,Koszinowski UH
14-0411 was used in Flow cytometry/Cell sorting to investigate the underlying mechanisms of extramedullary hematopoiesis.
Wed May 15 00:00:00 EDT 2013
Cathepsin G-dependent modulation of platelet thrombus formation in vivo by blood neutrophils.
PloS one
Faraday N,Schunke K,Saleem S,Fu J,Wang B,Zhang J,Morrell C,Dore S
Published figure using CD41a monoclonal antibody (Product # 14-0411-82) in Flow Cytometry
Mon Apr 14 00:00:00 EDT 2014
Cis-element mutated in GATA2-dependent immunodeficiency governs hematopoiesis and vascular integrity.
The Journal of clinical investigation
Johnson KD,Hsu AP,Ryu MJ,Wang J,Gao X,Boyer ME,Liu Y,Lee Y,Calvo KR,Keles S,Zhang J,Holland SM,Bresnick EH
Published figure using CD41a monoclonal antibody (Product # 14-0411-82) in Flow Cytometry
Mon Oct 01 00:00:00 EDT 2012
Etv2 is expressed in the yolk sac hematopoietic and endothelial progenitors and regulates Lmo2 gene expression.
Stem cells (Dayton, Ohio)
Koyano-Nakagawa N,Kweon J,Iacovino M,Shi X,Rasmussen TL,Borges L,Zirbes KM,Li T,Perlingeiro RC,Kyba M,Garry DJ
Published figure using CD41a monoclonal antibody (Product # 14-0411-82) in Flow Cytometry
Wed Aug 01 00:00:00 EDT 2012
Signaling profiling at the single-cell level identifies a distinct signaling signature in murine hematopoietic stem cells.
Stem cells (Dayton, Ohio)
Du J,Wang J,Kong G,Jiang J,Zhang J,Liu Y,Tong W,Zhang J
Published figure using CD41a monoclonal antibody (Product # 14-0411-82) in Flow Cytometry
Sun Jul 01 00:00:00 EDT 2012
Platelet activation attracts a subpopulation of effector monocytes to sites of Leishmania major infection.
The Journal of experimental medicine
Goncalves R,Zhang X,Cohen H,Debrabant A,Mosser DM
14-0411 was used in Immunofluorescence to point to a new role for platelets in host defense involving the selective recruitment of a subpopulation of effector monocytes from the blood to efficiently kill this intracellular parasite.
Mon Jun 06 00:00:00 EDT 2011
ER71 directs mesodermal fate decisions during embryogenesis.
Development (Cambridge, England)
Rasmussen TL,Kweon J,Diekmann MA,Belema-Bedada F,Song Q,Bowlin K,Shi X,Ferdous A,Li T,Kyba M,Metzger JM,Koyano-Nakagawa N,Garry DJ
Published figure using CD41a monoclonal antibody (Product # 14-0411-82) in Flow Cytometry
Tue Nov 01 00:00:00 EDT 2011
Acute systemic reaction and lung alterations induced by an antiplatelet integrin gpIIb/IIIa antibody in mice.
Blood
Nieswandt B,Echtenacher B,Wachs FP,Schröder J,Gessner JE,Schmidt RE,Grau GE,Männel DN
Thu Jul 15 00:00:00 EDT 1999
Acute systemic reaction and lung alterations induced by an antiplatelet integrin gpIIb/IIIa antibody in mice.
Blood
Nieswandt B,Echtenacher B,Wachs FP,Schröder J,Gessner JE,Schmidt RE,Grau GE,Männel DN
Thu Jul 15 00:00:00 EDT 1999
Acute systemic reaction and lung alterations induced by an antiplatelet integrin gpIIb/IIIa antibody in mice.
Blood
Nieswandt B,Echtenacher B,Wachs FP,Schröder J,Gessner JE,Schmidt RE,Grau GE,Männel DN
Thu Jul 15 00:00:00 EDT 1999
Acute systemic reaction and lung alterations induced by an antiplatelet integrin gpIIb/IIIa antibody in mice.
Blood
Nieswandt B,Echtenacher B,Wachs FP,Schröder J,Gessner JE,Schmidt RE,Grau GE,Männel DN
Thu Jul 15 00:00:00 EDT 1999