Search Thermo Fisher Scientific
Immunogen sequence: ASWGSPSAC PGAQAAPERL GRGVHDTDPW ATPPYCPKIQ SPPYSSQGTT SDASLWTPPQ GCPWTQSSPE PRNPPVPWTA APATLELAAV YQGLSVSPEP CLSLGAPSLL PHPSCQRLQP QTPGRCWSHS AEVVPNSEDQ GPGAAFQLSE ASPPQSSGLR FSGCPELWQE DLEGARLGIF Y (218-397 aa encoded by BC111413)
This gene encodes a member of the bHLH family of transcription factors and plays a key role in defining the rostrocaudal patterning of somites via interactions with multiple Notch signaling pathways. This gene is expressed in the anterior presomitic mesoderm and is downregulated immediately after the formation of segmented somites. This gene also plays a role in the formation of epithelial somitic mesoderm and cardiac mesoderm. Mutations in the MESP2 gene cause autosomal recessive spondylocostal dystosis 2 (SCD02).