Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
Mahogunin (MGRN1) is a C3HC4 RING-containing protein with E3 ubiquitin ligase activity in vitro.
Protein Aliases: flightless I actin binding protein; flightless I homolog; Protein flightless-1 homolog
Gene Aliases: FLII; FLIL