Based on 100% sequence identity, this antibody is predicted to react with Rat, Bovine and Orangutan
Protein phosphatase methylesterase-1 catalyzes the demethylation of the protein phosphatase-2A catalytic subunit.
Protein Aliases: PME-1; Protein phosphatase methylesterase 1; protein phosphatase methylesterase-1; testicular secretory protein Li 39
Gene Aliases: PME1; PP2593; PPME1; PRO0750