Invitrogen
This Antibody was verified by Cell treatment to ensure that the antibody binds to the antigen stated.
FIGURE: 1 / 6
ABS 043-29-02 binds free NGAL in mouse serum.
This antibody will not cross react with human, pig, dog, or rat.
ABS 043-29-02 may be used in Western blot and ELISA applications.
NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL
NGAL (Neutrophil gelatinase-associated lipocalin, lipocalin 2, siderocalin and neutrophil lipocalin) is a 25 kDa glycosylated single chain monomer and member of the lipocalin family of proteins that bind and transport small lipophilic molecules. NGAL is released by activated neutrophils, can form dimers and small amounts of higher oligomers, and complexes with matrix metalloproteinase 9 (MMP-9, gelatinase B). Low level expression of NGAL in a variety of epithelia may be increased in inflammation or cancers. Expression of NGAL in the kidney is dramatically increased by ischemic or nephrotoxic kidney injury.
Protein Aliases: 24p3; HGNC:6526; Lipocalin-2; Lipocalin2; Neutrophil gelatinase-associated lipocalin; NGAL; Oncogene 24p3; OTTHUMP00000022240; p25; RP23-161B9.11; secreted inducible protein 24; Siderocalin LCN2; SV-40-induced 24P3 protein; uterocalin
Gene Aliases: 24p3; AW212229; Lcn2; Sip24
UniProt ID: (Mouse) P11672
Entrez Gene ID: (Mouse) 16819
Molecular Function: transfer/carrier protein