Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MAQTPAFDKP KVELHVHLDG SIKPETILYY GRRRGIALPA NTAEGLLNVI GMDKPLTLPD FLAKFDYYMP AIAGCREAIK RIAYEFVEMK AKEGVVYVEV RYSPHLLANS KVEPIPWNQA EGDLTPDEVV ALVGQGLQEG ERDFGVKARS ILCCMRHQPN WSPKVVELCK KYQQQTVVAI DLAGDETIPG SSLLPGHVQA YQEAVKSGIH RTVHAGEVGS AEVVKEAVDI LKTERLGHGY HTLEDQALYN RLRQENMHFE ICPWSSYLTG AWKPDTEHAV IRLKNDQANY SLNTDDPLIF KSTLDTDYQM TKRDMGFTEE EFKRLNINAA KSSFLPEDEK RELLDLLYKA YGMPPSASAG QNL
Catalyzes the hydrolytic deamination of adenosine and 2-deoxyadenosine. Plays an important role in purine metabolism and in adenosine homeostasis. Modulates signaling by extracellular adenosine, and so contributes indirectly to cellular signaling events. May act as a positive regulator of T-cell coactivation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Adenosine aminohydrolase; Adenosine deaminase; RP11-61L14.5
Gene Aliases: ADA; ADA1
UniProt ID: (Human) P00813
Entrez Gene ID: (Human) 100
Molecular Function: deaminase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support