Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: RPLPEPLTVQ LLTVPGEVFP PKVKYTFFVP NDVDFSMQSS KERATTNIIQ PLLHAQWVLG DWSECSSTCG AGWQRRTVEC RDPSGQASAT CNKALKPEDA KPCESQLCPL
This gene encodes a member of the ADAMTS (a disintegrin and metalloproteinase with thrombospondin motifs) protein family. Members of the family share several distinct protein modules, including a propeptide region, a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 (TS) motif. Individual members of this family differ in the number of C-terminal TS motifs, and some have unique C-terminal domains. The enzyme encoded by this gene contains two C-terminal TS motifs, and disrupts angiogenesis in vivo. A number of disorders have been mapped in the vicinity of this gene, most notably lung neoplasms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: a disintegrin and metalloproteinase; A disintegrin and metalloproteinase with thrombospondin motifs 8; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 8; ADAM; ADAM metallopeptidase with thrombospondin type 1 motif, 8; ADAM-TS 8; ADAMs; ADAMTS-8; EC 3.4.24.14; EC 3.4.24.82; FLJ41712; metalloendopeptidases; METH-2; METH-8; METH2ADAM-TS METH-8
Gene Aliases: ADAM-TS8; ADAMTS8; METH2
UniProt ID: (Human) Q9UP79
Entrez Gene ID: (Human) 11095
Molecular Function: metalloprotease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support