Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MFAEIQIQDK DRMGTAGKVI KCKAAVLWEQ KQPFSIEEIE VAPPKTKEVR IKILATGICR TDDHVIKGTM VSKFPVIVGH EATGIVESIG EGVTTVKPGD KVIPLFLPQC RECNACRNPD GNLCIRSDIT GRGVLADGTT RFTCKGKPVH HFMNTSTFTE YTVVDESSVA KIDDAAPPEK VCLIGCGFST GYGAAVKTGK VKPGSTCVVF GLGGVGLSVI MGCKSAGASR IIGIDLNKDK FEKAMAVGAT ECISPKDSTK PISEVLSEMT GNNVGYTFEV IGHLETMIDA LASCHMNYGT SVVVGVPPSA KMLTYDPMLL FTGRTWKGCV FGGLKSRDDV PKLVTEFLAK KFDLDQLITH VLPFKKISEG FELLNSGQSI RTVLTF
This gene encodes class IV alcohol dehydrogenase 7 mu or sigma subunit, which is a member of the alcohol dehydrogenase family. Members of this family metabolize a wide variety of substrates, including ethanol, retinol, other aliphatic alcohols, hydroxysteroids, and lipid peroxidation products. The enzyme encoded by this gene is inefficient in ethanol oxidation, but is the most active as a retinol dehydrogenase; thus it may participate in the synthesis of retinoic acid, a hormone important for cellular differentiation. The expression of this gene is much more abundant in stomach than liver, thus differing from the other known gene family members.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alcohol dehydrogenase class 4 mu/sigma chain; Alcohol dehydrogenase class IV mu/sigma chain; alcohol dehydrogenase VII; alcohol dehydrogenase-7; All-trans-retinol dehydrogenase [NAD(+)] ADH7; class IV sigma-1 alcohol dehydrogenase; class IV sigmasigma alcohol dehydrogenase; Gastric alcohol dehydrogenase; Omega-hydroxydecanoate dehydrogenase ADH7; ret; Retinol dehydrogenase
Gene Aliases: ADH4; ADH7
UniProt ID: (Human) P40394
Entrez Gene ID: (Human) 131
Molecular Function: dehydrogenase oxidoreductase metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support