Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: VLGEVKVGVS YKLNGKRIPY FPANQEMLQK VEVEYETLPG WKADTTGARR WEDLPPQAQN YIRFVENH
ADSSL1 is a muscle isozyme of adenylosuccinate synthase (EC 6.3.4.4), which catalyzes the initial reaction in the conversion of inosine monophosphate (IMP) to adenosine monophosphate (AMP).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Adenylosuccinate synthetase isozyme 1; Adenylosuccinate synthetase, basic isozyme; Adenylosuccinate synthetase, muscle isozyme; Adenylosuccinate synthetase-like 1; adSS 1; AdSSL1; AMPSase 1; IMP--aspartate ligase 1; M-type adenylosuccinate synthetase
Gene Aliases: ADSS1; ADSSL1
UniProt ID: (Human) Q8N142
Entrez Gene ID: (Human) 122622
Molecular Function: ligase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support