Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: RGVFRRDAHK SEVAHRFKDL GEENFKALVL IAFAQYLQQC PFEDHVKLVN EVTEFAKTCV ADESAENCDK SLHTLFGDKL CTVATLRETY GEMADCCAKQ EPERNECFLQ HKDDNPNLPR LVRPEVDVMC TAFHDNEETF LKKYLYEIAR RHPYFYAPEL LFFAKRYKAA FTECCQAADK AACLLPKLDE LRDEGKASSA KQRLKCASLQ KFGERAFKAW AVARLSQRFP KAEFAEVSKL VTDLTKVHTE CCHGDLLECA DDRADLAKYI CENQDSISSK LKECCEKPLL EKSHCIAEVE NDEMPADLPS LAADFVESKD VCKNYAEAKD VFLGMFLYEY ARRHPDYSVV LLLRLAKTYE TTLEKCCAAA DPHECYAKVF DEFKPLVEEP QNLIKQNCEL FEQLGEYKFQ NALLVRYTKK VPQVSTPTLV EVSRNLGKVG SKCCKHPEAK RMPCAEDYLS VVLNQLCVLH EKTPVSDRVT KCCTESLVNR RPCFSALEVD ETYVPKEFNA ETFTFHADIC TLSEKERQIK KQTALVELVK HKPKATKEQL KAVMDDFAAF VEKCCKADDK ETCFAEGGKK LVAASQAALG L
Albumin is a soluble, monomeric protein which comprises about one-half of the blood serum protein. Albumin functions primarily as a carrier protein for steroids, fatty acids, and thyroid hormones and plays a role in stabilizing extracellular fluid volume. Albumin is a globular unglycosylated serum protein of molecular weight 65,000. Albumin is synthesized in the liver as preproalbumin which has an N-terminal peptide that is removed before the nascent protein is released from the rough endoplasmic reticulum. The product, proalbumin, is in turn cleaved in the Golgi vesicles to produce the secreted albumin.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Albumin; albumin (32 AA); albumin (AA 34); cell growth inhibiting protein 42; growth-inhibiting protein 20
Gene Aliases: ALB; ANALBA; FDAH; GIG20; GIG42; PRO0883; PRO0903; PRO1341; PRO1708; PRO2044; PRO2619; PRO2675; UNQ696/PRO1341
UniProt ID: (Human) Q8IUK7
Entrez Gene ID: (Human) 213
Molecular Function:
transfer/carrier protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support