Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: SEEDTLSLVT ADHSHVFSFG GYPLRGSSIF GLAPGKARDR KAYTVLLYGN GPGYVLKDGA RPDVTESESG SPEYRQQSAV PLDGETHAGE
There are at least four distinct but related alkaline phosphatases: intestinal, placental, placental-like, and liver/bone/kidney (tissue non-specific). The product of this gene is a membrane bound glycosylated enzyme, localized to testis, thymus.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alkaline phosphatase Nagao isozyme; Alkaline phosphatase, germ cell type; Alkaline phosphatase, placental-like; alkaline phosphatase, placental-like 2; ALP-1; GCAP; Germ cell alkaline phosphatase; Nagao isozyme; Placental alkaline phosphatase-like; placental-like alkaline phosphatase; PLAP-like; testicular and thymus alkaline phosphatase
Gene Aliases: ALPG; ALPPL; ALPPL2; GCAP
UniProt ID: (Human) P10696
Entrez Gene ID: (Human) 251
Molecular Function: phosphatase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support