Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: VCSTVQKPD AANYYYLIIL KAGAENMVAT PLASTSNSLN GDALTFTTTF TLQDVSNDFE INIEVYSLVQ KKDPSGLDKK KKTSKSKAIT PKRLLTSITT KSNIHSSVMA SPGGLSAVRT SNFALVGSYT LSLSSVGNTK FVLDKVPFLS SLEGHIYLKI KCQVNSSVEE RGFLTIFEDV SGFGAWHRRW CVLSGNCISY WTYPDDEKRK NPIGRINLAN CTSRQIEPAN REFCARRNTF ELITVRPQRE DDRETLVSQC RDTLCVTKNW LSADTKEERD LWMQKLNQVL VDIRLWQPDA CYKPIGKP (818-1124 aa encoded by BC034692)
This gene encodes an actin-binding protein that plays a role in cell growth and migration, and in cytokinesis. The encoded protein is thought to regulate actin cytoskeletal dynamics in podocytes, components of the glomerulus. Mutations in this gene are associated with focal segmental glomerulosclerosis 8. Alternative splicing results in multiple transcript variants encoding different isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: actin-binding protein anillin; Anillin; anillin, actin binding protein; anillin, actin binding protein (scraps homolog, Drosophila); DKFZp779A055
Gene Aliases: 1110037A17Rik; 2900037I21Rik; ANLN; C78101; FSGS8; RGD1561367; scra; Scraps
UniProt ID: (Human) Q9NQW6, (Mouse) Q8K298
Entrez Gene ID: (Human) 54443, (Rat) 363031, (Mouse) 68743
Molecular Function: non-motor actin binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support