Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
Apolipoprotein A4 (also known as ApoA-IV) is a plasma protein that is O-linked glycoprotein after proteolytic processing. ApoA4 is a major constituent of HDL and it also can be found on the surface of newly synthesized chylomicrons. The physiological function of ApoA4 is not completely identified so far, but several evidences from vitro studies indicate that this apolipoprotein involves in several steps of the reverse cholesterol transport pathway, which transports cholesterol from peripheral cells back to the liver.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Apo-AV; apoA-V; Apolipoprotein A-V; apolipoprotein A-V precursor variant 3; Apolipoprotein A5; apolipoprotein AV; FLJ97995; MGC126836; MGC126838; Regeneration-associated protein 3
Gene Aliases: 1300007O05Rik; APOA5; APOAV; RAP3; UNQ411/PRO773
UniProt ID: (Human) Q6Q788, (Mouse) Q8C7G5
Entrez Gene ID: (Human) 116519, (Mouse) 66113
Molecular Function:
apolipoprotein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support